siRNA Details
Gene Symbol: U2af2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205231.1 | NP_001192160.1 | 12 | 1520 | ||
NM_133671.3 | NP_598432.2 | 12 | 1508 | |||
GenBank mRNA | AK078496.1 | - | 12 | 1474 | ||
BC007487.1 | - | 8 | 1742 | 3593924 | ||
BC043071.1 | - | 12 | 1508 | 6403830 | ||
BC094451.1 | - | 10 | 1161 | |||
BC106134.1 | - | 11 | 1224 | 6309073 | ||
X64587.1 | - | 12 | 1426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
