siRNA Details
Gene Symbol: Ube2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019668.3 | NP_062642.1 | 5, 6 | 460 | ||
GenBank mRNA | AF089812.1 | - | 5, 6 | 468 | ||
AF383148.1 | - | 5, 6 | 468 | |||
AK150417.1 | - | 5, 6 | 443 | |||
AK153327.1 | - | 5, 6 | 443 | |||
AK160527.1 | - | 5, 6 | 455 | |||
AK160538.1 | - | 5, 6 | 469 | |||
AK160545.1 | - | 5, 6 | 467 | |||
AK163966.1 | - | 5, 6 | 486 | |||
AK164074.1 | - | 5, 6 | 469 | |||
BC026053.1 | - | 5, 6 | 458 | 4012075 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
