siRNA Details
Gene Symbol: Usp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009462.1 | NP_033488.1 | 11 | 2018 | ||
GenBank mRNA | AK077453.1 | - | 11 | 2040 | ||
AK129083.1 | - | 11 | 2026 | |||
AK153675.1 | - | 11 | 2510 | |||
AK157970.1 | - | 11 | 2042 | |||
AK165156.1 | - | 11 | 2038 | |||
AK166243.1 | - | 11 | 2034 | |||
AK172443.1 | - | 11 | 2040 | |||
AK202299.1 | - | 2 | 201 | |||
BC007134.1 | - | 11 | 2020 | 3491297 | ||
D84096.1 | - | 11 | 2018 |
