siRNA Details
Gene Symbol: Ucp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011671.5 | NP_035801.3 | 5 | 863 | ||
GenBank mRNA | AF111998.1 | - | 3 | 566 | ||
AF111999.1 | - | 3 | 560 | |||
AK035298.1 | - | 858 | ||||
AK054060.1 | - | 864 | ||||
AK134116.1 | - | 5 | 864 | |||
AK142687.1 | - | 5 | 864 | |||
AK165245.1 | - | 5 | 864 | |||
BC012697.1 | - | 5 | 862 | 4205625 | ||
BC012967.1 | - | 4 | 764 | 2811421 | ||
U69135.1 | - | 5 | 791 | |||
U94593.1 | - | 5 | 784 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
