siRNA Details
Gene Symbol: Umps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009471.2 | NP_033497.1 | 3 | 450 | ||
GenBank mRNA | AK035821.1 | - | 3 | 451 | ||
AK035997.1 | - | 2 | 297 | |||
AK090102.1 | - | 3 | 453 | |||
AK159808.1 | - | 3 | 451 | |||
AK159955.1 | - | 3 | 451 | |||
BC003887.1 | - | 3 | 447 | 3590810 | ||
BC095981.1 | - | 3 | 436 | 3499037 |
