siRNA Details
Gene Symbol: Nr1h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177730.1 | NP_001171201.1 | 3 | 263 | ||
NM_013839.4 | NP_038867.2 | 3 | 416 | |||
GenBank mRNA | AF085745.1 | - | 3 | 158 | ||
AJ132601.1 | - | 3 | 194 | |||
AK143904.1 | - | 3 | 197 | |||
AK159250.1 | - | 3 | 417 | |||
AY195870.1 | - | 3 | 263 | |||
AY195871.1 | - | 3 | 263 | |||
BC012646.1 | - | 3 | 204 | 4163109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
