siRNA Details
Gene Symbol: Uox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009474.5 | NP_033500.1 | 3, 4 | 414 | ||
GenBank mRNA | AK050047.1 | - | 2, 3 | 204 | ||
AK050425.1 | - | 3, 4 | 415 | |||
BC019771.1 | - | 3, 4 | 396 | 5136328 | ||
M27695.1 | - | 3, 4 | 369 |
