siRNA Details
Gene Symbol: Upk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009476.2 | NP_033502.2 | 3 | 393 | ||
GenBank mRNA | BC104260.1 | - | 3 | 393 | 40039813 | |
BC104261.1 | - | 3 | 393 | 40039814 | ||
DQ358978.1 | - | 3 | 332 | |||
U08030.1 | - | 3 | 391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
