siRNA Details
Gene Symbol: Uros | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA272198.1 | - | 4, 5 | 426 | ||
AK135314.1 | - | 5, 6 | 482 | |||
AK145902.1 | - | 5, 6 | 532 | |||
AK148116.1 | - | 5, 6 | 669 | |||
AK164297.1 | - | 5, 6 | 447 | |||
BC068161.1 | - | 5, 6 | 475 | 30358504 | ||
U04439.1 | - | 4, 5 | 396 | |||
U16216.1 | - | 5, 6 | 537 | |||
U18867.2 | - | 5, 6 | 541 |

More Information
Other Species Targeted

