siRNA Details
Gene Symbol: Uros | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135314.1 | - | 7 | 1159 | ||
AK145902.1 | - | 8 | 717 | |||
AK148116.1 | - | 8 | 854 | |||
BC068161.1 | - | 8 | 660 | 30358504 | ||
U04439.1 | - | 7 | 581 | |||
U16216.1 | - | 8 | 722 | |||
U18867.2 | - | 8 | 726 |
