siRNA Details
Gene Symbol: Scgb1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011681.2 | NP_035811.1 | 2 | 278 | ||
GenBank mRNA | AK145844.1 | - | 2 | 243 | ||
BC027518.1 | - | 2 | 235 | 1434396 | ||
CB595042.1 | - | 2 | 282 | |||
L04503.1 | - | 2 | 304 | |||
X67702.1 | - | 2 | 285 |
Gene Symbol: Bpifb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC089529.1 | - | 1040 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
