siRNA Details
Gene Symbol: Utrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011682.4 | NP_035812.3 | 36 | 5206 | ||
GenBank mRNA | AK087303.1 | - | 10 | 1865 | ||
Y12229.1 | - | 36 | 5114 |

More Information
Other Species Targeted

Gene Ontology Categories:
