siRNA Details
Gene Symbol: Slc45a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053077.3 | NP_444307.1 | 3 | 899 | ||
GenBank mRNA | AF360357.1 | - | 3 | 807 | ||
AK029155.1 | - | 3 | 900 | |||
AK089932.1 | - | 3 | 870 | |||
AK148207.1 | - | 897 | ||||
AY034377.1 | - | 3 | 820 | |||
BC125376.1 | - | 3 | 886 | 40129891 | ||
BC132429.1 | - | 3 | 886 | 40130706 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
