siRNA Details
Gene Symbol: Uxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013840.3 | NP_038868.2 | 3 | 200 | ||
GenBank mRNA | AF092738.1 | - | 3 | 187 | ||
AK027917.1 | - | 3 | 201 | |||
AK038495.1 | - | 2 | 743 | |||
AK086822.1 | - | 3 | 209 | |||
AK163983.1 | - | 3 | 240 | |||
BC029258.1 | - | 3 | 183 | 4483276 | ||
BC082294.1 | - | 2 | 169 | 30290324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
