siRNA Details
Gene Symbol: Vax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009501.1 | NP_033527.1 | 1 | 205 | ||
GenBank mRNA | AF064554.1 | - | 1 | 205 | ||
BC111818.1 | - | 1 | 204 | 40044698 | ||
BC119455.1 | - | 1 | 203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
