siRNA Details
Gene Symbol: Vcam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011693.3 | NP_035823.3 | 3 | 871 | ||
GenBank mRNA | AK016465.1 | - | 3 | 640 | ||
AK030195.1 | - | 3 | 641 | |||
AK143401.1 | - | 3 | 641 | |||
AK162954.1 | - | 3 | 641 | |||
BC011159.1 | - | 3 | 613 | 4206476 | ||
BC029823.1 | - | 3 | 871 | 4196681 | ||
CT010373.1 | - | 3 | 550 | |||
L08431.1 | - | 3 | 610 | |||
M84487.1 | - | 3 | 607 | |||
U12884.1 | - | 3 | 593 | |||
X67783.1 | - | 3 | 565 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
