siRNA Details
Gene Symbol: Vcam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC011159.1 | - | 2 | 175 | 4206476 | |
BC029823.1 | - | 2 | 433 | 4196681 | ||
CT010373.1 | - | 2 | 112 | |||
L08431.1 | - | 2 | 172 | |||
M84487.1 | - | 2 | 169 | |||
U12884.1 | - | 2 | 155 | |||
X67783.1 | - | 2 | 127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
