siRNA Details
Gene Symbol: Vdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011695.2 | NP_035825.1 | 8 | 764 | ||
GenBank mRNA | AK012359.1 | - | 8 | 762 | ||
AK078555.1 | - | 8 | 769 | |||
AK150504.1 | - | 8 | 790 | |||
AK150627.1 | - | 8 | 790 | |||
AK150940.1 | - | 8 | 782 | |||
AK152274.1 | - | 8 | 762 | |||
AK159561.1 | - | 8 | 999 | |||
AK168199.1 | - | 8 | 735 | |||
AK168328.1 | - | 8 | 761 | |||
BC003731.1 | - | 8 | 764 | 3601668 | ||
U30838.1 | - | 8 | 764 |

More Information
Other Species Targeted

Gene Ontology Categories:
