siRNA Details
Gene Symbol: Vegfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025250.3 | NP_001020421.2 | 3 | 1228 | ||
NM_001025257.3 | NP_001020428.2 | 3 | 1228 | |||
NM_001287056.1 | NP_001273985.1 | 3 | 1228 | |||
NM_001287057.1 | NP_001273986.1 | 3 | 1228 | |||
NM_001287058.1 | NP_001273987.1 | 3 | 1228 | |||
NM_009505.4 | NP_033531.3 | 3 | 1228 | |||
GenBank mRNA | AY120866.1 | - | 1 | 106 | ||
AY263146.1 | - | 3 | 207 | |||
AY707864.1 | - | 4 | 425 | |||
AY750956.1 | - | 4 | 386 | |||
AY750957.1 | - | 4 | 349 | |||
AY756068.1 | - | 3 | 164 | |||
BC061468.1 | - | 3 | 601 | 6816435 | ||
BQ832724.1 | - | 2 | 129 | |||
CA321456.1 | - | 3 | 601 | |||
EF028705.1 | - | 4 | 421 | |||
M95200.1 | - | 3 | 293 | |||
S37052.1 | - | 3 | 219 | |||
S38083.1 | - | 3 | 219 | |||
S38100.1 | - | 3 | 219 | |||
U50279.1 | - | 3 | 207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
