siRNA Details
Gene Symbol: Vegfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185164.1 | NP_001172093.1 | 3, 4 | 537 | ||
NM_011697.3 | NP_035827.1 | 3, 4 | 537 | |||
GenBank mRNA | AK148188.1 | - | 3, 4 | 536 | ||
AK178982.1 | - | 1, 2 | 168 | |||
AK207652.1 | - | 1, 2 | 164 | |||
AK208173.1 | - | 1, 2 | 168 | |||
AK211743.1 | - | 1, 2 | 169 | |||
AK216634.1 | - | 1, 2 | 168 | |||
BC046303.1 | - | 3, 4 | 515 | 6492277 | ||
U43836.1 | - | 3, 4 | 489 | |||
U43837.1 | - | 3, 4 | 297 | |||
U48800.1 | - | 3, 4 | 391 | |||
U52820.1 | - | 3, 4 | 391 |

More Information
Other Species Targeted

Gene Ontology Categories:
