siRNA Details
Gene Symbol: Lin7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011699.3 | NP_035829.1 | 2 | 64 | ||
GenBank mRNA | AF087695.1 | - | 2 | 64 | ||
AF173083.1 | - | 2 | 71 | |||
AK015399.1 | - | 2 | 83 | |||
AK078923.1 | - | 2 | 84 | |||
AK083299.1 | - | 2 | 87 | |||
AK128989.1 | - | 2 | 84 | |||
AK133531.1 | - | 2 | 73 | |||
AK135810.1 | - | 2 | 90 | |||
AK161076.1 | - | 2 | 45 | |||
BC046966.1 | - | 2 | 64 | 6493071 | ||
BC052471.1 | - | 2 | 74 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
