siRNA Details
Gene Symbol: Vipr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011703.4 | NP_035833.2 | 8 | 965 | ||
GenBank mRNA | AF266282.1 | - | 8 | 847 | ||
AK052465.1 | - | 8 | 966 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
