siRNA Details
Gene Symbol: Wap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011709.5 | NP_035839.3 | 2 | 241 | ||
GenBank mRNA | AK142655.1 | - | 242 | |||
AK164735.1 | - | 2 | 242 | |||
BC132280.1 | - | 2 | 239 | 40130557 | ||
BC132282.1 | - | 2 | 239 | 40130559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
