siRNA Details
Gene Symbol: ITIH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002216.2 | NP_002207.2 | 14, 15 | 1941 | ||
GenBank mRNA | AK225277.1 | - | 16, 17 | 1804 | ||
AK225329.1 | - | 14, 15 | 1846 | |||
AK291379.1 | - | 14, 15 | 1889 | |||
AK297334.1 | - | 9, 10 | 1132 | |||
BC132685.1 | - | 14, 15 | 1828 | 40146707 | ||
M18193.1 | - | 4, 5 | 632 | |||
M33033.1 | - | 4, 5 | 630 | |||
X07173.1 | - | 14, 15 | 1842 |
