siRNA Details
Gene Symbol: Ercc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011729.2 | NP_035859.2 | 8 | 2033 | ||
GenBank mRNA | AK137563.1 | - | 8 | 1865 | ||
AK140825.1 | - | 8 | 1862 | |||
AK160581.1 | - | 2 | 907 | |||
AK160645.1 | - | 6 | 1405 | |||
AK164205.1 | - | 8 | 1851 | |||
BC033413.1 | - | 8 | 1753 | |||
BC053034.1 | - | 8 | 1842 | |||
BC060659.1 | - | 8 | 1832 | |||
BC092235.1 | - | 8 | 1824 | |||
BC141070.1 | - | 8 | 1863 | 9055682 | ||
D16306.1 | - | 8 | 1728 | |||
U40796.1 | - | 8 | 1929 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
