siRNA Details
Gene Symbol: Slc6a20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011731.3 | NP_035861.2 | 11 | 1984 | ||
GenBank mRNA | AF075261.1 | - | 11 | 1808 | ||
AK076806.1 | - | 12 | 1715 | |||
AK143893.1 | - | 11 | 1957 | |||
AK144038.1 | - | 11 | 1985 | |||
AK163340.1 | - | 12 | 1873 | |||
AK164706.1 | - | 11 | 1829 | |||
AK164713.1 | - | 9 | 3861 | |||
BC013484.1 | - | 11 | 1895 | 4242168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
