siRNA Details
Gene Symbol: Yap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171147.1 | NP_001164618.1 | 2 | 712 | ||
NM_009534.3 | NP_033560.1 | 2 | 712 | |||
GenBank mRNA | AK157234.1 | - | 2 | 712 | ||
BC014733.1 | - | 2 | 709 | 4239820 | ||
BC039125.1 | - | 2 | 567 | 2649070 | ||
BC094313.1 | - | 2 | 700 | 5697882 | ||
X80508.1 | - | 2 | 704 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
