siRNA Details
Gene Symbol: Zfp101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009542.2 | NP_033568.2 | 4 | 1414 | ||
GenBank mRNA | AK076355.1 | - | 4 | 1418 | ||
AK076461.1 | - | 1 | 1204 | |||
AK078458.1 | - | 4 | 1402 | |||
AK169234.1 | - | 4 | 1415 | |||
U07861.1 | - | 4 | 1374 |

More Information
Other Species Targeted

Gene Ontology Categories:
