siRNA Details
Gene Symbol: Sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110791.1 | NP_001104261.1 | 5 | 804 | ||
NM_011750.2 | NP_035880.2 | 5 | 804 | |||
GenBank mRNA | AK133895.1 | - | 5 | 593 | ||
AK146151.1 | - | 5 | 565 | |||
AK147081.1 | - | 5 | 573 | |||
AK152914.1 | - | 4 | 506 | |||
AK154705.1 | - | 5 | 573 | |||
AK157845.1 | - | 5 | 573 | |||
BC009091.1 | - | 5 | 549 | 3157495 | ||
BC055370.1 | - | 5 | 563 | 30055109 | ||
X85802.1 | - | 5 | 561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
