siRNA Details
Gene Symbol: Sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110791.1 | NP_001104261.1 | 6, 7 | 1058 | ||
NM_011750.2 | NP_035880.2 | 6, 7 | 1058 | |||
GenBank mRNA | AK133895.1 | - | 6, 7 | 847 | ||
AK146151.1 | - | 6, 7 | 819 | |||
AK147081.1 | - | 6, 7 | 827 | |||
AK152914.1 | - | 5, 6 | 760 | |||
AK154705.1 | - | 6, 7 | 826 | |||
AK157845.1 | - | 6, 7 | 827 | |||
BC009091.1 | - | 6, 7 | 803 | 3157495 | ||
BC055370.1 | - | 6, 7 | 817 | 30055109 | ||
X85802.1 | - | 6, 7 | 815 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
