siRNA Details
Gene Symbol: Zfp185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109043.1 | NP_001102513.1 | 15 | 1361 | ||
NM_009549.3 | NP_033575.3 | 15 | 1364 | |||
GenBank mRNA | AK014497.1 | - | 4 | 412 | ||
AK048188.1 | - | 15 | 1342 | |||
BC039772.1 | - | 12 | 949 | 4191053 | ||
EF494744.1 | - | 15 | 1344 | |||
U46687.1 | - | 14 | 1190 |
