siRNA Details
Gene Symbol: Zfand5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009551.5 | NP_033577.1 | 6 | 1037 | ||
GenBank mRNA | AF062071.1 | - | 5 | 629 | ||
AK076397.1 | - | 6 | 1038 | |||
AK131911.1 | - | 6 | 1033 | |||
AK216435.1 | - | 3 | 214 | |||
BC048232.1 | - | 4 | 1410 | |||
BC061693.1 | - | 2 | 3781 | |||
BC080270.1 | - | 6 | 1018 | |||
BC107566.1 | - | 6 | 601 | 5389770 | ||
BC119124.1 | - | 6 | 1038 | 8733873 | ||
BC119126.1 | - | 6 | 1038 | 8733875 | ||
L37047.1 | - | 1 | 68 |
