siRNA Details
Gene Symbol: Zfp54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011760.2 | NP_035890.2 | 4 | 1008 | ||
GenBank mRNA | AF080070.1 | - | 4 | 969 | ||
AK148292.1 | - | 4 | 1001 | |||
BC080782.1 | - | 5 | 1060 | 30468186 |
