siRNA Details
Gene Symbol: Zfp54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011760.2 | NP_035890.2 | 4 | 1801 | ||
GenBank mRNA | AF080070.1 | - | 4 | 1762 | ||
AK148292.1 | - | 4 | 1794 | |||
BC080782.1 | - | 5 | 1853 | 30468186 |
