siRNA Details
Gene Symbol: Zfp57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013745.2 | NP_001013767.1 | 5 | 1430 | ||
NM_001168501.1 | NP_001161973.1 | 5 | 1296 | |||
NM_001168502.1 | NP_001161974.1 | 6 | 1672 | |||
GenBank mRNA | AK054083.1 | - | 5 | 1413 | ||
AK135881.1 | - | 5 | 1404 | |||
AK136241.1 | - | 5 | 1414 | |||
AK139625.1 | - | 6 | 1672 | |||
AK143118.1 | - | 3 | 2278 | |||
AK145368.1 | - | 5 | 1402 | |||
BC052028.1 | - | 3 | 1597 | 5686931 | ||
D21850.1 | - | 4 | 1425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
