siRNA Details
Gene Symbol: Zfp85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001130.2 | NP_001001130.1 | 4 | 1353 | ||
GenBank mRNA | BC055337.1 | - | 4 | 1353 | 6413269 | |
BC055756.1 | - | 4 | 1353 | 6413269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
