siRNA Details
Gene Symbol: Zfp94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199321.1 | NP_001186250.1 | 6 | 1048 | ||
NM_009568.3 | NP_033594.1 | 5 | 1009 | |||
GenBank mRNA | AK138205.1 | - | 6 | 989 | ||
BC005437.1 | - | 5 | 1009 | 3591835 | ||
BC095945.1 | - | 4 | 829 | 4936327 | ||
U46187.1 | - | 5 | 952 |

More Information
Other Species Targeted

