siRNA Details
Gene Symbol: Zfpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011766.5 | NP_035896.1 | 6, 7 | 1237 | ||
GenBank mRNA | AF107306.1 | - | 6, 7 | 758 | ||
AF118845.1 | - | 6, 7 | 1007 | |||
AF125166.1 | - | 6, 7 | 1238 | |||
BC059241.1 | - | 6, 7 | 1137 | 6408975 | ||
EF514219.1 | - | 6, 7 | 2060 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001130501.
Gene Ontology Categories:
