siRNA Details
Gene Symbol: Zfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011767.2 | NP_035897.2 | 10 | 1865 | ||
GenBank mRNA | AF071059.1 | - | 10 | 1863 | ||
AK034963.1 | - | 3 | 434 | |||
AK158938.1 | - | 2 | 173 | |||
BC094220.1 | - | 9 | 1752 | |||
BC141139.1 | - | 10 | 1849 | 9055751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
