siRNA Details
Gene Symbol: Zfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011767.2 | NP_035897.2 | 3, 4 | 488 | ||
GenBank mRNA | AF071059.1 | - | 3, 4 | 486 | ||
BC038599.1 | - | 3, 4 | 492 | |||
BC058570.1 | - | 3, 4 | 488 | |||
BC094220.1 | - | 2, 3 | 376 | |||
BC141139.1 | - | 3, 4 | 472 | 9055751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
