siRNA Details
Gene Symbol: Adam23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177600.1 | NP_001171071.1 | 7 | 1069 | ||
NM_011780.2 | NP_035910.1 | 7 | 1069 | |||
GenBank mRNA | AB009673.1 | - | 7 | 841 | ||
AK029301.1 | - | 5 | 384 | |||
AK034022.1 | - | 7 | 1064 | |||
AK046677.1 | - | 7 | 1083 |
