siRNA Details
Gene Symbol: Adam23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177600.1 | NP_001171071.1 | 19 | 2071 | ||
NM_011780.2 | NP_035910.1 | 19 | 2071 | |||
GenBank mRNA | AB009673.1 | - | 19 | 1843 | ||
AK029301.1 | - | 17 | 1386 | |||
AK034022.1 | - | 19 | 2066 | |||
AK046677.1 | - | 19 | 2085 |
