siRNA Details
Gene Symbol: Agr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011783.2 | NP_035913.1 | 3 | 227 | ||
GenBank mRNA | AB016592.1 | - | 3 | 226 | ||
AF044262.1 | - | 3 | 227 | |||
AK007677.1 | - | 3 | 246 | |||
BC013334.1 | - | 3 | 245 | 4218549 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
