siRNA Details
Gene Symbol: Aloxe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011786.2 | NP_035916.2 | 9 | 1254 | ||
GenBank mRNA | AK141003.1 | - | 8 | 4020 | ||
BC116258.1 | - | 9 | 1253 | 40093733 | ||
BC116259.1 | - | 9 | 1253 | 40093735 | ||
Y14695.1 | - | 9 | 1254 |

More Information
Other Species Targeted

Gene Ontology Categories:
