siRNA Details
Gene Symbol: Arih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011790.4 | NP_035920.1 | 12 | 1306 | ||
GenBank mRNA | AF124664.1 | - | 11 | 1133 | ||
AJ130975.1 | - | 11 | 1165 | |||
AK011094.1 | - | 12 | 1313 | |||
AK045012.1 | - | 1289 | ||||
AK149936.1 | - | 12 | 1286 | |||
AK167104.1 | - | 12 | 1263 | |||
AK168055.1 | - | 12 | 1311 | |||
BC051998.1 | - | 12 | 1306 | 6400287 | ||
BC052422.1 | - | 12 | 1306 | 6400287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
