siRNA Details
Gene Symbol: Banf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038231.2 | NP_001033320.1 | 3 | 624 | ||
NM_001286608.1 | NP_001273537.1 | 3 | 966 | |||
NM_011793.3 | NP_035923.1 | 3 | 832 | |||
GenBank mRNA | AB025349.1 | - | 3 | 564 | ||
AK003025.1 | - | 3 | 608 | |||
AK004212.1 | - | 3 | 591 | |||
AK010257.1 | - | 3 | 610 | |||
AK011079.1 | - | 3 | 613 | |||
AK011100.1 | - | 3 | 611 | |||
AK012588.1 | - | 3 | 594 | |||
AK088896.1 | - | 3 | 588 | |||
AK168039.1 | - | 3 | 613 | |||
AK185753.1 | - | 1 | 266 | |||
BC003709.1 | - | 3 | 601 | 3599828 | ||
CK780850.1 | - | 3 | 596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
