siRNA Details
Gene Symbol: Cd52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013706.2 | NP_038734.1 | 2 | 184 | ||
GenBank mRNA | AK088161.1 | - | 2 | 151 | ||
AK155728.1 | - | 2 | 185 | |||
BC027495.1 | - | 2 | 134 | 1396480 | ||
M55561.1 | - | 2 | 173 | |||
X76696.1 | - | 2 | 160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
