siRNA Details
Gene Symbol: Cdc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025779.1 | NP_001020950.1 | 7 | 1069 | ||
NM_011799.2 | NP_035929.1 | 7 | 1145 | |||
GenBank mRNA | AJ009559.1 | - | 7 | 1162 | ||
AJ223087.1 | - | 7 | 1147 | |||
AK012192.1 | - | 7 | 1099 | |||
AK075975.1 | - | 7 | 1097 | |||
AK085020.1 | - | 7 | 1099 | |||
AK162669.1 | - | 7 | 1107 | |||
AK166003.1 | - | 7 | 1150 | |||
BC052434.1 | - | 7 | 1069 | 6837113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
