siRNA Details
Gene Symbol: Clec5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038604.1 | NP_001033693.1 | 5 | 478 | ||
NM_021364.2 | NP_067339.1 | 4 | 403 | |||
GenBank mRNA | AF139769.1 | - | 4 | 373 | ||
AK036697.1 | - | 4 | 369 | |||
AK046600.1 | - | 4 | 390 | |||
AK137482.1 | - | 4 | 404 | |||
BC104364.1 | - | 5 | 437 | 40049817 | ||
BC104365.1 | - | 5 | 437 | 40049818 | ||
CN670295.1 | - | 5 | 440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
