siRNA Details
Gene Symbol: Def6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027185.3 | NP_081461.2 | 11 | 1853 | ||
GenBank mRNA | AF329497.1 | - | 11 | 1788 | ||
AK010356.1 | - | 3 | 327 | |||
AK038050.1 | - | 11 | 1824 | |||
AK088460.1 | - | 11 | 1802 | |||
AY241695.1 | - | 11 | 1788 | |||
AY278770.1 | - | 5 | 669 | |||
BC120500.1 | - | 11 | 1853 | 40129423 | ||
BC131953.1 | - | 11 | 1853 | 40130230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
